Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cbl-c Antibody (OTI4E3), PE/Cy7, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271756PECY7

Catalog No. N271756PEC7


Add to Cart

Description

Description

Cbl-c Monoclonal specifically detects Cbl-c in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cbl-c
Monoclonal
PE-Cyanine7
PBS with 0.05% Sodium Azide
CBLC
Recombinant protein fragment corresponding to amino acids 244-474 of human Cbl-c (NP_036248) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Flow Cytometry
OTI4E3
Flow Cytometry
Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C
Mouse
Affinity Purified
Stem Cell Markers
23624.0
Store at 4°C in the dark. Do not freeze.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit