Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerebellin 3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549564

Catalog No. PA549564

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in synaptic functions in the CNS.
Specifications

Specifications

Cerebellin 3
Polyclonal
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q6UW01, Q9JHG0
Cerebellin-3, CBLN3
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56410, 643866
Immunohistochemistry, Western Blot
Unconjugated
CBLN3
Liquid
CBLN3
synthetic peptide corresponding to 145-170aa of human CBLN3
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit