Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Calmodulin binding peptide Recombinant Rabbit Monoclonal Antibody (HL1841)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547197
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
CBP affinity tag is a useful tag in protein study which can be added to the N or C terminus of proteins of interest through DNA recombinant technology. The tag is derived from muscle myosin light-chain kinase. The tag comprises 26 amino acid residues with the molecular weight of 4 kDa and sequence of KRRWKKNFIAVSAANRFKKISSSGAL. CBP tag has the relatively high affinity for calmodulin (CaM). CBP-tagged proteins can be purified from crude cell extracts through CaM affinity resin. CBP Tag antibody is a useful tool in analysis of CBP-tagged proteins.
Specifications
Calmodulin binding peptide | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Tag | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
HL1841 | |
Unconjugated | |
AW558298; CBP; CBP/p300; CREB binding protein; CREB-binding protein; CREBBP; KAT3A; p300/CBP; RSTS; RTS | |
Synthetic peptide of human CBP tag. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction