Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBP tag Antibody (HL1841) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325400
Description
CBP tag Monoclonal antibody specifically detects CBP tag in Epitope Tag samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
CBP tag | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Tag | |
Purified |
Western Blot, Immunocytochemistry | |
HL1841 | |
Western Blot 1:1000-1:10000, Immunocytochemistry/ Immunofluorescence | |
26 aa C-terminal fragment from muscle myosin light-chain kinase, Calmodulin Binding Protein Epitope tag, Calmodulin Binding Protein tag | |
Synthtic peptide of human CBP tag. | |
100 μL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction