Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CBR 5884
3-Phosphoglycerate dehydrogenase (PHGDH) inhibitor
Supplier: Tocris Bioscience™ 5836/10
This item is not returnable.
View return policy
Description
CBR 5884 is a phosphoglycerate dehydrogenase (PHGDH) inhibitor (IC50 = 33 μM). Decreases de novo serine synthesis in cancer cells. Inhibits proliferation of cancer cell lines having high PHGDH expression levels.Specifications
Ethyl 5-[(2-furanylcarbonyl)amino]-3-methyl-4-thiocyanato-2-thiophenecarboxylate | |
10 mg | |
C14H12N2O4S2 | |
>98% |
681159-27-3 | |
Dehydrogenase Inhibitors | |
336.39 |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction