Learn More
Tocris Bioscience™ CBR 5884
3-Phosphoglycerate dehydrogenase (PHGDH) inhibitor
$166.20 - $694.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
583610 View Documents Tocris Bioscience™
5836/10
Each for $166.20
Add to cart
583650 View Documents Tocris Bioscience™
5836/50
Each for $694.00
Add to cart
Description
Description
Phosphoglycerate dehydrogenase (PHGDH) inhibitor (IC50 = 33 μM). Decreases de novo serine synthesis in cancer cells. Inhibits proliferation of cancer cell lines having high PHGDH expression levels.