Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBR1 Mouse anti-Human, Clone: OTI4B4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527219

Catalog No. PIMA527219


Add to cart

Description

Description

Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q.
Specifications

Specifications

CBR1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P16152
15 hydroxyprostaglandin dehydrogenase [NADP ];15-hydroxyprostaglandin dehydrogenase [NADP ];Carbonyl reductase [NADPH] 1;CBR 1;CBR1;CBR1_HUMAN;CRN;NADPH dependent carbonyl reductase 1;NADPH-dependent carbonyl reductase 1;Prostaglandin 9 ketoreductase; Prostaglandin 9-ketoreductase;Prostaglandin E(2) 9 reductase;Prostaglandin-E(2) 9-reductase;SDR21C1.
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
873
Immunohistochemistry (Paraffin), Western Blot
OTI4B4
Unconjugated
CBR1
Liquid
CBR1
Full length human recombit protein of CBR1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit