Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBR1 Mouse anti-Human, Clone:, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524765

Catalog No. PIMA524765

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q.
Specifications

Specifications

CBR1
Monoclonal
PBS with 50% glycerol and 0.02% sodium azide
P16152
15 hydroxyprostaglandin dehydrogenase [NADP ];15-hydroxyprostaglandin dehydrogenase [NADP ];Carbonyl reductase [NADPH] 1;CBR 1;CBR1;CBR1_HUMAN;CRN;NADPH dependent carbonyl reductase 1;NADPH-dependent carbonyl reductase 1;Prostaglandin 9 ketoreductase; Prostaglandin 9-ketoreductase;Prostaglandin E(2) 9 reductase;Prostaglandin-E(2) 9-reductase;SDR21C1.
Mouse
IgG1
100 μL
-20°C
Primary
873
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CBR1
Liquid
CBR1
Human CBR1 recombit protein
Purified
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit