Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBR3 Mouse anti-Human, Mouse, Rat, Canine, Monkey, Biotin, Clone: OTI1G6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270343B
Description
CBR3 Monoclonal antibody specifically detects CBR3 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CBR3 | |
OTI1G6 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
CBR3 | |
Full length recombinant protein of human CBR3 (NP_001227) produced in HEK293T cells. | |
Immunogen affinity purified | |
Lipid and Metabolism | |
Primary | |
874.0 |
Immunohistochemistry, Western Blot | |
Biotin | |
PBS with 0.05% Sodium Azide | |
carbonyl reductase (NADPH) 3, carbonyl reductase (NADPH) 3, EC 1.1.1.18410EC 1.1.1, carbonyl reductase [NADPH] 3, carbonyl reductase 3, EC 1.1.1.184, hCBR3, NADPH-dependent carbonyl reductase 3, SDR21C2, short chain dehydrogenase/reductase family 21C, member 2 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Canine, Human, Monkey, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title