Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBR3 Mouse anti-Human, Mouse, Rat, Canine, Monkey, Janelia Fluor™ 646, Clone: OTI1G6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270343JF646

Catalog No. N270343J646


Add to cart

Description

Description

CBR3 Monoclonal antibody specifically detects CBR3 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Immunohistochemistry.
Specifications

Specifications

CBR3
OTI1G6
Immunohistochemistry
Affinity Purified, Conjugated
CBR3
Full length recombinant protein of human CBR3 (NP_001227) produced in HEK293T cells.
Immunogen affinity purified
Lipid and Metabolism
Primary
874.0
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
carbonyl reductase (NADPH) 3, carbonyl reductase (NADPH) 3, EC 1.1.1.18410EC 1.1.1, carbonyl reductase [NADPH] 3, carbonyl reductase 3, EC 1.1.1.184, hCBR3, NADPH-dependent carbonyl reductase 3, SDR21C2, short chain dehydrogenase/reductase family 21C, member 2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit