Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBR3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548471

Catalog No. PA548471

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Carbonyl reductase 3 catalyzes the reduction of a large number of biologically and pharmacologically active carbonyl compounds to their corresponding alcohols. The enzyme is classified as a monomeric NADPH-dependent oxidoreductase. CBR3 contains three exons spanning 11.2 kilobases and is closely linked to another carbonyl reductase gene - CBR1.
Specifications

Specifications

CBR3
Polyclonal
PBS with 0.09% sodium azide
O75828
Carbonyl reductase [NADPH] 3, NADPH-dependent carbonyl reductase 3, CBR3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
874
Flow Cytometry, Western Blot
Unconjugated
CBR3
Liquid
CBR3
KLH conjugated synthetic peptide between 195-224 amino acids from the C-terminal region of human CBR3.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit