Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS Mouse anti-Human, Mouse, Rat, Alexa Fluor™ 594, Clone: 519, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243604AF594

Catalog No. NP243604A94


Add to cart

Description

Description

CBS Monoclonal antibody specifically detects CBS in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications

Specifications

CBS
519
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
cystathionine-beta-synthase, EC 4.2.1.2210Beta-thionase
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CBS
Recombinant protein encompassing a sequence within the center region of human CBS. The exact sequence is proprietary.
0.1 mL
Primary
875
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit