Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: 519, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243604UV

Catalog No. NBP243604UV


Add to cart

Description

Description

CBS Monoclonal antibody specifically detects CBS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications

Specifications

CBS
519
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated
Purified
CBS
Recombinant protein encompassing a sequence within the center region of human CBS. The exact sequence is proprietary.
Protein G purified
Lipid and Metabolism
Primary
875
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
cystathionine-beta-synthase, EC 4.2.1.2210Beta-thionase
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit