Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 147871AP150UL
Description
Strongly expressed in human liver and pancreas, with weaker expression in heart and brain, the cytoplasmic protein cystathionine b-synthase (CBS) operates in the first step of homocysteine transulfuration. CBS, which belongs to the cysteine synthase/cystathionine b-synthase family of proteins, catalyzes the formation of cystathionine from the thrombogenic amino acid homocysteine using pyridoxal phosphate cofactor. Allosteric activation by adenosyl-methionine regulates CBS activity. Deficiencies in CBS are assoc-iated with homocystinuria, a recessively inherited error in sulfur amino acid metabolism that affects many organs and tissues. Symptoms of homocytinuria include arteriosclerosis, thrombosis, dislocated optic lenses, mental retard-ation and skeletal abnormalities.Specifications
CBS | |
Polyclonal | |
Unconjugated | |
CBS | |
Beta thionase, CBS, cystathionine beta synthase, HIP4, Serine sulfhydrase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12411, 24250, 875 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32232, P35520, Q91WT9 | |
CBS | |
CBS Fusion Protein Ag6437 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction