Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBWD1 Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549319

Catalog No. PA549319


Add to cart

Specifications

Specifications

Cbwd1
Polyclonal
PBS with 0.09% sodium azide
Q99MB4
COBW domain-containing protein 1, Cobalamin synthase W domain-containing protein 1, Cbwd1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
171057
Western Blot
Unconjugated
Cbwd1
Liquid
CBWD1
KLH conjugated synthetic peptide between 325-351 amino acids from the C-terminal region of rat Cbwd1.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit