Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen CBX1
Concentration 0.25 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-6141
Encompass_Preferred
View Documents
Proteintech Group IncSupplier Diversity Partner
102412AP150UL
150 μL
Each for $395.30
Add to cart
 
Description

Description

This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family. The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The protein may play an important role in the epigenetic control of chromatin structure and gene expression. Several related pseudogenes are located on chromosomes 1, 3, and X. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications

Specifications

CBX1
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Polyclonal
-20°C
0.25 mg/mL
Unconjugated
Rabbit
Human, Mouse, Rat
IgG
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit