Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CBX2 Monoclonal Antibody (4D10B2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548796
Description
CBX2 Monoclonal Antibody for Western Blot, ICC/IF, IHC, Flow, ELISA
MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
Specifications
CBX2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P30658, Q14781 | |
CBX2 | |
Purified recombinant fragment of human CBX2 (AA: 402-525) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
4D10B2 | |
Unconjugated | |
CBX2 | |
Cbx2; CDCA6; cell division cycle associated 6; chromobox 2; chromobox homolog 2; chromobox homolog 2 (Pc class homolog, Drosophila); chromobox protein homolog 2; hromobox homolog 2; M33; M33 polycomb-like protein; MOD2; modifier 3; modifier 3 protein; pc; Pc class homolog; polycomb; SRXY5 | |
Mouse | |
Protein G | |
RUO | |
12416, 84733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction