Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBX4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP310881100UL

 View more versions of this product

Catalog No. NB126663


Add to cart

Description

Description

CBX4 Polyclonal specifically detects CBX4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CBX4
Unconjugated
PBS buffer, 2% sucrose
chromobox homolog 4, chromobox homolog 4 (Drosophila Pc class), Chromobox protein homolog 4, Drosophila), E3 SUMO-protein ligase CBX4, hPC2, NS5ATP1-binding protein 16, Pc class 2 homolog, PC2, Polycomb 2 homolog
The immunogen is a synthetic peptide directed towards the N terminal region of human CBX4 (NP_003646). Peptide sequence LLIAFQNRERQEQLMGYRKRGPKPKPLVVQVPTFARRSNVLTGLQDSSTD
100 μg
Apoptosis
Primary
8535
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit