Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBX7 Mouse anti-Human, Clone: 3H9C4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261723

Catalog No. NBP2-61723


Add to cart

Description

Description

CBX7 Monoclonal antibody specifically detects CBX7 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

CBX7
3H9C4
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Purified
CBX7
Purified recombinant fragment of human CBX7 (AA: 15-147) expressed in E. Coli.
93 kDa
0.1 ml
Primary
23492
ELISA, Western Blot
Unconjugated
PBS with 0.05% Sodium Azide
chromobox homolog 7, chromobox protein homolog 7
Mouse
IgG1
Protein G purified
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit