Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cbx8 Mouse anti-Human, Biotin, Clone: 9C3D4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237411B

Catalog No. NBP237411B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Cbx8
9C3D4
Western Blot, ELISA
Purified
CBX8
Purified recombinant fragment of human CBX8 (AA: 17-222) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Western Blot
Biotin
PBS with 0.05% Sodium Azide
chromobox homolog 8, chromobox homolog 8 (Drosophila Pc class), chromobox homolog 8 (Pc class homolog, Drosophila), hPc3, Pc3, PC3chromobox protein homolog 8, polycomb 3, Polycomb 3 homolog, RC1Pc class 3 homolog, Rectachrome 1
Mouse
IgG1
0.1 ml
Primary
57332
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit