Learn More
Azzota Corp CC-115 DFS Item
Manufacturer:  Azzota Corp C1154
CC-115 is a dual inhibitor of DNA-dependent protein kinase (DNA-PK) and mammalian target of rapamycin (mTOR), with potential antineoplastic activity. Please request if need more quantities
Catalog No. 50-161-3117