Learn More
CC Chemokine Receptor D6 Rat anti-Human, Alexa Fluor™ 594, Clone: 25G14, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221998AF594
Catalog No. NP221998A94
Add to cart
Description
Description
CC Chemokine Receptor D6 Monoclonal antibody specifically detects CC Chemokine Receptor D6 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CC Chemokine Receptor D6
25G14
Flow Cytometry
C-C chemokine receptor D6, CCR10CCR9D6CC-chemokine-binding receptor JAB61, chemokine (C-C) receptor 9, chemokine binding protein 2, Chemokine receptor CCR-10, Chemokine receptor CCR-9, chemokine receptor D6, chemokine-binding protein 2, Chemokine-binding protein D6, CMKBR9chemokine (C-C motif) receptor 9, hD6, MGC126678, MGC138250
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
ACKR2
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human CC chemokine receptor D6. Rat anti human CC Chemokine Receptor D6 O00590.
0.1 mL
Primary
1238