Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CC Chemokine Receptor D6 Rat anti-Human, PE/Cy5.5, Clone: 25G14, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP221998PECY55
Description
CC Chemokine Receptor D6 Monoclonal antibody specifically detects CC Chemokine Receptor D6 in Human samples. It is validated for Flow Cytometry.Specifications
CC Chemokine Receptor D6 | |
25G14 | |
Flow Cytometry | |
Conjugated, Purified | |
ACKR2 | |
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human CC chemokine receptor D6. Rat anti human CC Chemokine Receptor D6 O00590. | |
Protein G purified | |
Autophagy, Cellular Markers, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
Primary | |
1238.0 |
Flow Cytometry | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
C-C chemokine receptor D6, CCR10CCR9D6CC-chemokine-binding receptor JAB61, chemokine (C-C) receptor 9, chemokine binding protein 2, Chemokine receptor CCR-10, Chemokine receptor CCR-9, chemokine receptor D6, chemokine-binding protein 2, Chemokine-binding protein D6, CMKBR9chemokine (C-C motif) receptor 9, hD6, MGC126678, MGC138250 | |
Rat | |
IgG2 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title