Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CC2D1A Rabbit anti-Human, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

Supplier:  Abnova Corporation PAB30390

Catalog No. 89-032-734


Add to Cart

Description

Description

Rabbit polyclonal antibody raised against partial recombinant human CC2D1A.

  • High specificity
  • High reactivity
Specifications

Specifications

CC2D1A
Polyclonal
Rabbit polyclonal antibody raised against partial recombinant human CC2D1A.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
FLJ20241/FLJ41160/FREUD-1/Freud-1/Aki1/MRT3
Rabbit
Antigen affinity purification
RUO
54862
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) The optimal working dilution should be determined by the end user.
Q6P1N0
CC2D1A
Recombinant protein corresponding to human CC2D1A.
100 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)