Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

CCBL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549033

Catalog No. PA549033

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a cytosolic enzyme that is responsible for the metabolism of cysteine conjugates of certain halogenated alkenes and alkanes. This metabolism can form reactive metabolites leading to nephrotoxicity and neurotoxicity. Increased levels of this enzyme have been linked to schizophrenia. Multiple transcript variants that encode different isoforms have been identified for this gene.
Specifications

Specifications

CCBL1
Polyclonal
PBS with 0.09% sodium azide
Q16773
Kynurenine--oxoglutarate transaminase 1, Cysteine-S-conjugate beta-lyase, Glutamine transaminase K, GTK, Glutamine--phenylpyruvate transaminase, Kynurenine aminotransferase I, KATI, Kynurenine--oxoglutarate transaminase I, CCBL1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
883
Western Blot
Unconjugated
KYAT1
Liquid
KYAT1
KLH conjugated synthetic peptide between 327-355 amino acids from the C-terminal region of human CCBL1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit