Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC105 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532078
Description
Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH. Predicted reactivity: Chimpanzee (99%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Specifications
CCDC105 | |
Polyclonal | |
Unconjugated | |
CCDC105 | |
CCDC105; coiled-coil domain containing 105; coiled-coil domain-containing protein 105 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126402 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IYK2 | |
CCDC105 | |
Recombinant fragment corresponding to a region within amino acids 142 and 412 of Human CCDC105. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction