Learn More
Abnova™ CCDC108 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00255101T02
Catalog No. 89-938-233
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CCDC108 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 18.5kDa
Transient overexpression cell lysate was tested with anti-CCDC108 antibody (Mfr. No. H00255101-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
BC031585
pCMV-CCDC108 full-length
100 μL
Human