Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC110 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548270

Catalog No. PA548270


Add to cart

Specifications

Specifications

CCDC110
Polyclonal
PBS with 0.09% sodium azide
Q8TBZ0, Q3V125
Coiled-coil domain-containing protein 110, Cancer/testis antigen 52, CT52, Cancer/testis antigen KM-HN-1, CCDC110, KMHN1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
212392, 256309
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CCDC110
Liquid
CCDC110
KLH conjugated synthetic peptide between 777-806 amino acids from the C-terminal region of human CCDC110.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit