Learn More
CCDC112 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29989
Catalog No. 89-033-277
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human CCDC112.
  • High specificity
  • High reactivity
Specifications
Specifications
CCDC112
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (1.25 ug/mL) The optimal working dilution should be determined by the end user.
NM_001040440
MBC1|MGC39633
Rabbit
Protein A purification
RUO
Primary
153733
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human CCDC112.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
CCDC112
A synthetic peptide corresponding to N-terminus of human CCDC112.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)