Learn More
CCDC12 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 251381AP
Catalog No. 50-173-4745
Add to cart
Description
Description
The exact function of this protein remains unknown.
Specifications
Specifications
CCDC12
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WUD4
CCDC12
Rabbit
IgG
150 μL
-20°C
Primary
151903
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
CCDC12
Liquid
CCDC12
CCDC12 Fusion Protein Ag18223
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human