Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC122 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565914
Description
Immunogen sequence: GEVESKWSFMT ELHEKRDFVK KLKTMKEELM QDLQNPGGNR ITQVQEDITN LKDKIITVKE SIIE Highest antigen sequence identity to the following orthologs - mouse 62%, rat 65%.
This gene encodes a protein that contains a coiled-coil domain. Naturally occurring mutations in this gene are associated with leprosy.
Specifications
CCDC122 | |
Polyclonal | |
Unconjugated | |
CCDC122 | |
CCDC122; coiled-coil domain containing 122; coiled-coil domain-containing protein 122 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
160857 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T0U0 | |
CCDC122 | |
Recombinant Human CCDC122. Recombinant protein control fragment (Product #RP-106591). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction