Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548421

Catalog No. PA548421

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Required for primary cilia formation and promotes the localization of the ciliopathy protein BBS4 to both centriolar satellites and cilia.
Specifications

Specifications

CCDC13
Polyclonal
PBS with 0.09% sodium azide
Q8IYE1
Coiled-coil domain-containing protein 13, CCDC13
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
152206
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CCDC13
Liquid
CCDC13
KLH conjugated synthetic peptide between 51-79 amino acids from the N-terminal region of human CCDC13.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit