Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC135 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | CCDC135 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB126333
|
Novus Biologicals
NBP310716100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
CCDC135 Polyclonal specifically detects CCDC135 in Human samples. It is validated for Western Blot.Specifications
CCDC135 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
C16orf50, chromosome 16 open reading frame 50, coiled-coil domain containing 135, coiled-coil domain-containing protein 135, DKFZp434I099 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human CCDC135 (NP_115645). Peptide sequence ELEVLEILKLREEEEAAHTLTISIYDTKRNEKSKEYREAMERMMHEEHLR | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
84229 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title