Learn More
Aviva Systems Biology Corp CCDC138 Antibody - N-terminal region (ARP52772_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52772_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC138. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCDC138. Alternate names: FLJ32745ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60828