Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC144C Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549087

Catalog No. PA549087


Add to cart

Specifications

Specifications

CCDC144C
Polyclonal
PBS with 0.09% sodium azide
Q8IYA2
Putative coiled-coil domain-containing protein 144C, Coiled-coil domain-containing protein 144C pseudogene, CCDC144CP, CCDC144C
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
348254
Western Blot
Unconjugated
CCDC144CP
Liquid
CCDC144CP
KLH conjugated synthetic peptide between 1064-1090 amino acids from the C-terminal region of human CCDC144C.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit