Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC144NL Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | CCDC144NL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157054
|
Novus Biologicals
NBP157054 |
100 μL |
Each of 1 for $436.00
|
|
Description
CCDC144NL Polyclonal specifically detects CCDC144NL in Human samples. It is validated for Western Blot.Specifications
CCDC144NL | |
Polyclonal | |
Rabbit | |
Human | |
Q6NUI1 | |
339184 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
coiled-coil domain containing 144 family, N-terminal like, MGC87631, putative coiled-coil domain-containing protein 144 N-terminal-like | |
CCDC144NL | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title