Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC147 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557714
Description
Immunogen sequence: EKGGKQVLEE SAFEEMERDF QGVLHELSGD KSLEKFRIEY ERLHAVMKKS YDNEKRLMAK CRELNAEIVV NSAKVATALK LSQDDQTTIA SLKKE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
CFAP58 is a protein coding gene.
Specifications
CCDC147 | |
Polyclonal | |
Unconjugated | |
CFAP58 | |
bA127L20.4; bA127L20.4 (novel protein); bA127L20.5; bA127L20.5 (novel protein); bA554P13.1; C10orf80; CCDC147; Cfap58; cilia and flagella associated protein 58; cilia- and flagella-associated protein 58; coiled-coil domain containing 147; coiled-coil domain-containing protein 147; coiled-coil domain-containing protein 147-like; Gm969 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
159686 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T655 | |
CFAP58 | |
Recombinant protein corresponding to Human CCDC147. Recombinant protein control fragment (Product #RP-97072). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction