Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC151 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156716
Description
CCDC151 Polyclonal specifically detects CCDC151 in Human samples. It is validated for Western Blot.Specifications
CCDC151 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
coiled-coil domain containing 151, coiled-coil domain-containing protein 151, FLJ31801, MGC20983 | |
Rabbit | |
Affinity purified | |
RUO | |
115948 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
A5D8V7 | |
CCDC151 | |
Synthetic peptides corresponding to MGC20983 The peptide sequence was selected from the middle region of MGC20983. Peptide sequence IALPLATSKDKFFDEESEEEDNEVVTRASLKIRSQKLIESHKKHRRSRRS. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Crab-eating macaque: 92%;. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction