Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC157 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | CCDC157 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17048820
|
Novus Biologicals
NBP17048820UL |
20 μL |
Each for $152.22
|
|
NBP170488
|
Novus Biologicals
NBP170488 |
100 μL |
Each for $436.00
|
|
Description
CCDC157 Polyclonal specifically detects CCDC157 in Human samples. It is validated for Western Blot.Specifications
CCDC157 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
550631 | |
Synthetic peptides corresponding to LOC550631(hypothetical LOC550631) The peptide sequence was selected from the middle region of LOC550631. Peptide sequence DKEKARVDSMVRHQESLQAKQRALLKQLDSLDQEREELRGSLDEAEAQRA. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
coiled-coil domain containing 157, KIAA1656 | |
CCDC157 | |
IgG | |
Affinity Purified | |
83 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title