Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC19 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550886

Catalog No. PA550886


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total CFAP45 protein.

The function remains unknown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCDC19
Polyclonal
Unconjugated
CFAP45
1700028D05Rik; CCDC19; CFAP45; cilia and flagella associated protein 45; cilia- and flagella-associated protein 45; coiled-coil domain containing 19; Coiled-coil domain-containing protein 19; coiled-coil domain-containing protein 19, mitochondrial; nasopharyngeal epithelium specific protein 1; nasopharyngeal epithelium specific protein 1 (NESG1); nasopharyngeal epithelium-specific protein 1; NESG1; protein CFAP45, mitochondrial; Unknown (protein for MGC:134208)
Rabbit
Antigen Affinity Chromatography
RUO
25790
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.5 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9UL16
CFAP45
Synthetic peptide corresponding to residues near the N terminal of human cilia and flagella associated protein 45.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.