Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC27 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA540115

Catalog No. PA540115


Add to cart

Description

Description

Recommended positive controls: K562, THP-1, and HL-60 cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

CCDC27
Polyclonal
Unconjugated
CCDC27
Liquid
CCDC27
Recombit protein encompassing a sequence within the C-terminus region of human CCDC27.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.71 mg/mL
0.1M tris with 0.1M glycine, 20% glycerol and 0.01% Thimerosal; pH 7
Q2M243
coiled-coil domain containing 27, RP1-286D6.1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
148870
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit