Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC27 Mouse anti-Human, Clone: OTI1F8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527416

Catalog No. PIMA527416


Add to cart

Description

Description

The function of this protein remains unknown.
Specifications

Specifications

CCDC27
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q2M243
coiled-coil domain containing 27, RP1-286D6.1
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
148870
Western Blot
OTI1F8
Unconjugated
CCDC27
Liquid
CCDC27
Full length human recombit protein of CCDC27 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit