Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC28A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548994

Catalog No. PA548994

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is located in a region close to the locus of the pseudogene of chemokine (C-C motif) receptor-like 1 on chromosome 6. The specific function of this gene has not yet been determined. [provided by RefSeq, Jul 2008]
Specifications

Specifications

CCDC28A
Polyclonal
PBS with 0.09% sodium azide
Q8IWP9
Coiled-coil domain-containing protein 28A, CCRL1AP, CCDC28A, C6orf80
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25901
Western Blot
Unconjugated
CCDC28A
Liquid
CCDC28A
KLH conjugated synthetic peptide between 79-107 amino acids from the N-terminal region of human CCDC28A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit