Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC28B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2846060.1ML
Description
CCDC28B Polyclonal specifically detects CCDC28B in Human samples. It is validated for Western Blot.Specifications
CCDC28B | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79140 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
coiled-coil domain containing 28B, coiled-coil domain-containing protein 28B, MGC1203, MGC16441, RP4-622L5.5 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human CCDC28B | |
0.1 mL | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction