Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC33 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | CCDC33 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB126517
|
Novus Biologicals
NBP310808100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
CCDC33 Polyclonal specifically detects CCDC33 in Human samples. It is validated for Western Blot.Specifications
CCDC33 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
cancer/testis antigen 61, coiled-coil domain containing 33, coiled-coil domain-containing protein 33, CT61, FLJ23168, FLJ32855, MGC34145 | |
The immunogen is a synthetic peptide directed towards the middle terminal region of human CCDC33 (NP_079331.3). Peptide sequence KVIEKMERVLEDRLQDRSKPPPLNRQQGKPYTGFPMLSASGLPLGSMGEN | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
80125 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title