Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC36 Monoclonal Antibody (OTI2D11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526980
Description
CCDC36 Monoclonal Antibody for Western Blot, IHC (P)
Required for DNA double-strand breaks (DSBs) formation in unsynapsed regions during meiotic recombination. Probably acts by forming a complex with MEI4 and REC114, which activates DSBs formation in unsynapsed regions, an essential step to ensure completion of synapsis. Not required for HORMAD1 functions in pairing-independent synaptonemal complex formation, ATR recruitment to unsynapsed axes, meiotic silencing of unsynapsed chromatin (MSUC) or meiotic surveillance.
Specifications
CCDC36 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IYA8 | |
CCDC36 | |
Full length human recombinant protein of CCDC36 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2D11 | |
Unconjugated | |
CCDC36 | |
Cancer/testis antigen 74; Ccdc36; coiled-coil domain containing 36; coiled-coil domain-containing protein 36; CT74; IHO1; Interactor of HORMAD1 protein 1 | |
Mouse | |
Affinity chromatography | |
RUO | |
339834 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction