Learn More
CCDC36 Mouse anti-Human, Clone: OTI5E11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808463
Catalog No. 50-166-8876
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Required for DNA double-strand breaks (DSBs) formation in unsynapsed regions during meiotic recombination. Probably acts by forming a complex with MEI4 and REC114, which activates DSBs formation in unsynapsed regions, an essential step to ensure completion of synapsis. Not required for HORMAD1 functions in pairing-independent synaptonemal complex formation, ATR recruitment to unsynapsed axes, meiotic silencing of unsynapsed chromatin (MSUC) or meiotic surveillance.
Specifications
Specifications
CCDC36
Monoclonal
Unconjugated
CCDC36
Lyophilized
CCDC36
Full length human recombit protein of human CCDC36 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5E11
PBS with 8% trehalose and no preservative; pH 7.3
Q8IYA8
CT74
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
339834