Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC48 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546240
Description
Peptide sequence: LEAELQRYRS EDSQLPTPQL ANPEPGDKSN EPEDAGTRDP DPTPEGAWQS Sequence homology: Human: 100%; Rat: 77%.
The function of this protein remains unknown.
Specifications
CCDC48 | |
Polyclonal | |
Unconjugated | |
EFCC1 | |
C3orf73; Ccdc48; Ccdc48l1; coiled-coil domain containing 48; coiled-coil domain containing 48 like; coiled-coil domain containing 48-like1; coiled-coil domain-containing protein 48; EFCC1; EF-hand and coiled-coil domain containing 1; EF-hand and coiled-coil domain-containing protein 1; EF-hand domain-containing protein ENSP00000381169; EF-hand domain-containing protein ENSP00000381169 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79825 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9HA90 | |
EFCC1 | |
Synthetic peptide directed towards the C-terminal of human CCDC48 (aa 319-368). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction