Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC51 Mouse anti-Human, Clone: A8-F4-D1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267221
Description
CCDC51 Monoclonal antibody specifically detects CCDC51 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCDC51 | |
A8-F4-D1 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
coiled-coil domain containing 51, coiled-coil domain-containing protein 51, FLJ12436 | |
Mouse | |
IgG1 | |
100μL | |
Primary | |
79714 |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin | |
Purified | |
CCDC51 | |
Recombinant protein with human CCDC51 C terminal | |
Protein A purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title