Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSRP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548505

Catalog No. PA548505

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

RNA-binding protein that mediates pre-mRNA alternative splicing regulation.
Specifications

Specifications

NSRP1
Polyclonal
PBS with 0.09% sodium azide
Q9H0G5
Nuclear speckle splicing regulatory protein 1, Coiled-coil domain-containing protein 55, Nuclear speckle-related protein 70, NSrp70, NSRP1, CCDC55, NSRP70
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84081
Western Blot
Unconjugated
NSRP1
Liquid
NSRP1
KLH conjugated synthetic peptide between 92-121 amino acids from the N-terminal region of human CCDC55.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit